Beschreibung
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
Spezifikation
Spezifikation
| Antigen | MED20 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.18 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | MED20 |
| Gen-Zugriffsnummer | Q5XIE9, Q9H944, Q9R0X0 |
| Gen-Alias | hTRFP, MED20, mediator complex subunit 20, PRO0213, TRF proximal protein homolog, TRFP |
| Gensymbole | Med20 |
| Mehr anzeigen |