missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Menin Recombinant Rabbit Monoclonal Antibody (8G11)
Alle ansehen Thermo Scientific Produkte

Beschreibung
This antibody has been tested in direct-ELISA.
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
Spezifikation
Spezifikation
| Antigen | Menin |
| Anwendungen | Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Klon | 8G11 |
| Konzentration | 0.58 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol, 150 mM NaCl and 0.02% sodium azide; pH 7.4 |
| Gen | MEN1 |
| Gen-Zugriffsnummer | O00255 |
| Gen-Alias | AW045611; MEAI; MEN1; Menin; menin 1; multiple endocrine neoplasia 1; multiple endocrine neoplasia I; SCG2 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?