Learn More
Mer Antibody (125518) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals FAB8912B
Artikelnummer. 18620244
  • ONLINE EXCLUSIVE PRICE
    710.00 €
    670.95 € / 0.10 Milliliter
    Sparen 39.05 €
    5% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Mer Monoclonal antibody specifically detects Mer in Human samples. It is validated for Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF, Gene Knock-Out
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Mer
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF, Gene Knock-Out
125518
Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready, Knockout Validated
c-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer, Met1-Ala499, Accession # AAB60430
0.1 mL
Cancer, Protein Kinase, Signal Transduction, Vision
10461
Store at 4°C in the dark.
IgG2b
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen