missing translation for 'onlineSavingsMsg'
Learn More

Mesothelin Antibody (MB-G10), PE, Novus Biologicals™

Artikelnummer. 18515684 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18515684 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18515684 Supplier Novus Biologicals Supplier No. NB11085538PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mesothelin Monoclonal antibody specifically detects Mesothelin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mesothelin
Anwendungen Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon MB-G10
Konjugat PE
Zusammensetzung PBS
Gen-Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPFSMRP, Pre-pro-megakaryocyte-potentiating factor, soluble MPF mesothelin related protein
Wirtsspezies Mouse
Immunogen This Mesothelin Antibody (MB-G10) was produced in mesothelin-deficient mice by immunizations with plasmid cDNA encoding human Mesothelin full length protein followed by a single boost of a recombinant human mesothelin-Fc fusion protein. (Uniprot: Q13421)
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Cancer
Primär oder sekundär Primary
Gen-ID (Entrez) 10232
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.