missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metadherin Antibody (CL0397) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52929PCP
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Metadherin Monoclonal antibody specifically detects Metadherin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Tekniske data
| Metadherin | |
| Monoclonal | |
| PerCP | |
| 3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC | |
| This antibody was developed using a recombinant protein derived from Q86UE4, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Phospho Specific | |
| 92140 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0397 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion