missing translation for 'onlineSavingsMsg'
Learn More

Methyl-1-Testosteron, TRC

Product Code. 30475810
Change view
Click to view available options
Menge:
25 mg
Unit Size:
25 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30475810 25 mg 25 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475810 Supplier Toronto Research Chemicals Supplier No. TRCM32641025MG
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 17b-Hydroxy-17a-methyl-5a-androst-1-ene-3-one
CAS 65-04-3
Hinweise zur Reinheitsqualität HPLC
Summenformel C20H30O2
Menge 25 mg
Synonym 17β-Hydroxy-17α-methyl-5α-androst-1-ene-3-one, Methyl 1-Testosterone, 5α-Androst-1-en-3-one, 17β-hydroxy-17-methyl- (6CI,7CI,8CI), 17-Methyl-1-testosterone, 17α-Methyl-17β-hydroxy-5α-androst-1-en-3-one, 17α-Methyl-17β-hydroxy-Δ1-5α-androsten-3-one, 17β-Hydroxy-17-methyl-5α-androst-1-en-3-one, 17β-Hydroxy-17α-methyl-5α-androst-1-en-3-one, 17β-Hydroxy-17α-methyl-5α-androstan-1-en-3-one, Methyl-1-testosterone, SC 11195
InChI-Formel InChI=1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,13,15-17,22H,4-5,7-8,10-12H2,1-3H3/t13-,15+,16-,17-,18-,19-,20-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-Name (5S,8R,9S,10R,13S,14S,17S)-17-Hydroxy-10,13,17-trimethyl-5,6,7,8,9,11,12,14,15,16-decahydro-4H-cyclopenta[a]phenanthren-3-on
Molekulargewicht (g/mol) 302.4
Formelmasse 302.451
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.