missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-4-Butyrylamino-3-methylbenzoat, Mikromol™
Beschreibung
Methyl-4-Butyrylamino-3-methylbenzoat
Spezifikation
Spezifikation
| Chemischer Name oder Material | Methyl 4-Butyrylamino-3-methylbenzoate |
| CAS | 301533-59-5 |
| Bezeichnung der Analyten oder Bestandteile | Methyl 4-Butyrylamino-3-methylbenzoate |
| Versandbedingung | Room Temperature |
| Summenformel | C13 H17 N O3 |
| Zertifizierung/Konformität | ISO 9001 |
| Synonym | Benzoic acid, 3-methyl-4-[(1-oxobutyl)amino]-, methyl ester, 3-Methyl-4-butyrylaminobenzoic acid methyl ester, 4-Butyrylamino-3-methylbenzoic acid methyl ester, Methyl 4-Butyrylamino-3-methylbenzoate |
| InChI-Formel | InChI=1S/C13H17NO3/c1-4-5-12(15)14-11-7-6-10(8-9(11)2)13(16)17-3/h6-8H,4-5H2,1-3H3,(H,14,15) |
| SMILES | CCCC(=O)Nc1ccc(cc1C)C(=O)OC |
| IUPAC-Name | Methyl-4-(butanoylamino)-3-methylbenzoat |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?