missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Parathion-d6 (dimethyl-d6), CDN
Click to view available options
Menge:
0.01 g
0.05 g
Packungsgröße:
0.01 Gramm
0.05 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Parathion-methylD6(dimethylD6) |
| CAS | 96740-32-8 |
| Summenformel | C8 2H6 H4 N O5 P S |
| Menge | 0.05 g |
| Synonym | Parathion-methyl D6 (dimethyl D6), Phosphorothioic acid, O,O-di(methyl-d3) O-(4-nitrophenyl) ester (9 CI) |
| InChI-Formel | InChI=1 S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])OP(=S)(Oc1ccc(cc1)[N+](=O)[O-])OC([2 H])([2 H])[2 H] |
| IUPAC-Name | (4-nitrophenoxy)-sulfanylidene-bis(trideuteriomethoxy)-λ^{5}-phosphane |
| Molekulargewicht (g/mol) | 269.24 |
| Formelmasse | 269.0394 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction