missing translation for 'onlineSavingsMsg'
Learn More

Methyl(S)-Pyroglutamat, TRC

Product Code. 30466330
Change view
Click to view available options
Menge:
1 g
2 g
5 g
Unit Size:
1 Gramm
2 Gramm
5 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30466330 2 g 2 Gramm
30465033 5 g 5 Gramm
30470916 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466330 Supplier Toronto Research Chemicals Supplier No. TRCM3259482G
 Eingeschränkter Artikel
Verfügbar ab: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4931-66-2
Summenformel C6 H9 N O3
Molekulargewicht (g/mol) 143.14
IUPAC-Name Methyl-(2S)-5-oxopyrrolidin-2-carboxylat
SMILES COC(=O)[C@@H]1CCC(=O)N1

Specifications

Chemischer Name oder Material Methyl (S)-Pyroglutamate
Menge 2 g
InChI-Formel InChI=1S/C6H9NO3/c1-10-6(9)4-2-3-5(8)7-4/h4H,2-3H2,1H3,(H,7,8)/t4-/m0/s1
Formelmasse 143.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.