Learn More
Metriram (> 90%), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Metiram |
| CAS | 9006-42-2 |
| Summenformel | (C16H33N11S16Zn3)x |
| Menge | 1 g |
| Synonym | Metiram, Polyram, Amarex, Carbatene, Metiram-zinc, NIA 9102, Phytox-Super, Polycarbacin, Polycarbacine, Polycarbazin, Polycarbazine, Polykarbacin, Polymarcin, Polymarcine, Polymarsin, Polymarzin, Polymarzine, Polymat, Polyram 2000, Polyram 80, Polyram 80WP, Polyram DF, Polyram combi |
| InChI-Formel | InChI=1S/C4H8N2S4.Zn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2 |
| SMILES | C(CNC(=S)[S-])NC(=S)[S-].[Zn+2] |
| IUPAC-Name | Zink(II)-ethan-1,2-diyldicarbamodithioat |