Learn More
MFN1 Mouse anti-Human, CoraLite 488, Clone: 3F11C11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66776-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The MFN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MFN1. This antibody recognizes human antigen. The MFN1 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Spezifikation
| MFN1 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry | |
| 3F11C11 | |
| CoraLite 488 | |
| 100 μL | |
| 55669 | |
| IgG2a |