missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage (′GnT-VI′ activity).
Spezifikation
Spezifikation
| Antigen | MGAT4C |
| Anwendungen | Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.19 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | MGAT4C |
| Gen-Zugriffsnummer | Q9UBM8 |
| Gen-Alias | GlcNAc T IVc, GnT IVc, GNTIVH, HGNT IV H, MGAT4C |
| Gensymbole | Mgat4c |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?