Learn More
Invitrogen™ MGST1 Recombinant Rabbit Monoclonal Antibody (24GB5110)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557078
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MGST1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.
Spezifikation
| MGST1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MGST1 | |
| 1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4257, 56615 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5110 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10620, Q91VS7 | |
| MGST1 | |
| A synthesized peptide derived from human MGST1 (aa 20-55). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |