Learn More
Invitrogen™ MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), PerCP-eFluor™ 710, eBioscience™

Mouse Monoclonal Antibody
304.00 €
Spezifikation
| Antigen | MHC Class I (H-2Kd/H-2Dd) |
|---|---|
| Klon | 34-1-2S |
| Konzentration | 0.2 mg/mL |
| Inhalt und Lagerung | 4°C, store in dark, DO NOT FREEZE! |
| Anwendungen | Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15519336
![]() |
Invitrogen™
46-5998-82 |
100 μg |
304.00 €
100 Mikrogramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p. Applications Reported: This 34-1-2S antibody has been reported for use in flow cytometric analysis. Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirical...
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spezifikation
| MHC Class I (H-2Kd/H-2Dd) | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01895, P01896, P01897, P01899, P01900, P01901, P03991, P04223, P14426, P14427, P14428 | |
| 14964, 14972 | |
| Primary | |
| Affinity chromatography |
| 34-1-2S | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Mouse | |
| H2-D1 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, D-B alpha chain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2K1; H2-K1; H2KD; histocompatibility 2, D region locus 1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-D-q alpha-chain; MHC H2-K transplantation antigen; Q5k protein | |
| H2-D1, H2-K1 | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.