Learn More
Invitrogen™ MHC I Free Chain Monoclonal Antibody (A4), eBioscience™, Invitrogen™

Beschreibung
Description: The A4 monoclonal antibody reacts with the class I human leukocyte antigen (HLA) complex in the absence of beta2 microglobulin. There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with beta2 microglobulin (beta2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. Applications Reported: This A4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This A4 antibody has been tested by flow cytometric analysis of 3-day PHA-activated normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Spezifikation
Spezifikation
| Antigen | MHC I Free Chain |
| Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Klassifikation | Monoclonal |
| Klon | A4 |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.09% sodium azide; pH 7.2 |
| Gen-Alias | MHC Free |
| Wirtsspezies | Mouse |
| Reinigungsverfahren | Affinity chromatography |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.