Learn More
Invitrogen™ Milatuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558880
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Milatuzumab has been used in trials studying the treatment of Lupus Nephritis, Multiple Myeloma (MM), GVHD (Acute or Chronic), Myelodysplastic Syndrome, and Chronic Lymphocytic Leukemia, among others.
Spezifikation
| Milatuzumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CD74-DOX (ADC); hLL1; hLL1-DOX (ADC); MEDI-115 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |