Learn More
MLL3 Antibody, Novus Biologicals™
Artikelnummer. 18206502 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
100 μL
25 μL
Packungsgröße:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18206502 100 μL 100 Mikroliter
18674088 25 μL 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18206502 Lieferant Novus Biologicals Lieferanten-Nr. NBP257730
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
MLL3 Polyclonal specifically detects MLL3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen MLL3
Anwendungen Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Zusammensetzung PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gen-Alias ALR-like protein, DKFZp686C08112, EC 2.1.1.43, FLJ12625, HALRMGC119851, histone-lysine N-methyltransferase MLL3, histone-lysine N-methyltransferase, H3 lysine-4 specific, Homologous to ALR protein, KIAA1506FLJ38309, KMT2C, KMT2CMGC119852, Lysine N-methyltransferase 2C, MGC119853, myeloid/lymphoid or mixed-lineage leukemia 3, Myeloid/lymphoid or mixed-lineage leukemia protein 3
Gensymbole KMT2C
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TPPTMSQPTFPMVPQQLQHQQHTTVISGHTSPVRMPSLPGWQPNSAPAHLPLNPPRIQPPIAQLPIKTCTPAPGTVSNANPQ
Reinigungsverfahren Affinity Purified
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 58508
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG