missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ MMP1 Monoclonal Antibody (K29028_6B4), Invitrogen™

Artikelnummer. p-200088857
Skift visning
Klik for at se tilgængelige muligheder
Menge:
50 μL
200 μL
Pakningsstørrelse:
200 Mikrogramm
50 Mikrogramm
Artikelnummer. Menge unitSize
30195295 200 μL 200 Mikrogramm
30207993 50 μL 50 Mikrogramm
2 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30195295

Brand: Invitrogen™ MA556182

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QTHLTYRIEN YTPDLPRADV DHAIEKAFQL WSNVTPLTFT KVSEGQADIM ISFVRGDHRD NSPFDGPGGN LAHAFQPGPG IGGDAHFDED ERWTNNFREY NLHRVAAHEL GHSLGLSHST DIGALMYPSY TFSGDVQLAQ DDIDGIQAIY GRSQNPVQPI GPQTPKACDS KLTFDAITTI

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen MMP1
Anwendungen Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon K29028_6B4
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gen MMP1
Gen-Zugriffsnummer P03956
Gen-Alias 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1a; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1a; MMPs
Gensymbole MMP1
Wirtsspezies Mouse
Immunogen Recombinant human MMP1 (immunogen range is 111-290).
Reinigungsverfahren Protein A
Menge 200 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 4312
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG2a
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.