missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
MMP26 Polyclonal antibody specifically detects MMP26 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | MMP26 |
| Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Zusammensetzung | 0.2 um filtered solution in PBS |
| Gen-Alias | EC 3.4.24.-, Endometase, matrilysin 2, matrilysin-2, matrix metallopeptidase 26, matrix metalloproteinase 26, matrix metalloproteinase-26, MGC126590, MGC126592, MMP-26 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human MMP26 (Accession#: NP_068573.2; Thr90-Pro261) |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?