missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP8 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5149489
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
The MMP8 gene encodes matrix metalloproteinase-8, also known as collagenase-2, an enzyme belonging to the matrix metalloproteinase (MMP) family, which plays a pivotal role in the breakdown of extracellular matrix (ECM) components. MMP8 is primarily involved in the degradation of type I and III collagen, facilitating tissue remodeling and repair processes. This enzyme is produced by neutrophils and is active in areas of tissue inflammation and injury, where it modulates the ECM to aid in wound healing. MMP8 is implicated in various physiological processes, including embryonic development, angiogenesis, and bone remodeling. However, excessive or unregulated activity of MMP8 can lead to pathological conditions such as arthritis, periodontal disease, and cancer metastasis, where ECM degradation contributes to disease progression. Research into MMP8 aims to understand its regulation and involvement in disease states, providing insights into potential therapeutic strategies for targeting MMP activity in inflammatory and degenerative conditions.
Spezifikation
| MMP8 | |
| Polyclonal | |
| Unconjugated | |
| MMP8 | |
| BB138268; CLG1; Collagenase; Collagenase 2; collagenase-2; HNC; matrix metallo protease; matrix metallopeptidase 8; matrix metallopeptidase 8 (neutrophil collagenase); matrix metalloproteinase 8; matrix metalloproteinase 8 (neutrophil collagenase); Matrix metalloproteinase8; matrix metalloproteinase-8; MMP; MMP 8; Mmp8; MMP-8; MMPs; Neutrophil collagenase; neutrophil collagenease; PMN leukocyte collagenase; PMNL CL; PMNL collagenase; PMNLCL; PMNL-CL | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P22894 | |
| MMP8 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-8, Phe21-Gly467. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur