Learn More
Invitrogen™ Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate
Beschreibung
Lectin GS-II is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or Β-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The red-fluorescent lectin GS-II Alexa Fluor 594 conjugate can be used in fluorescence microscopy.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
Spezifikation
| Menge | 500 μg |
| Quelle | Africa Shrub legume |
| Konjugat | Alexa Fluor 594 |
| Inhalt und Lagerung | Store in freezer (-5 to -30°C) and protect from light. |
| Markertyp | Alexa Fluor Dyes |
| Versandbedingung | Room Temperature |
| Form | Lyophilized |
| Protein-Subtyp | Lectins |
| Produktlinie | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.