missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus A29L (MPXV/USA/2022/MA001) Recombinant Rabbit Monoclonal Antibody (HL2527)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556794
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Monkeypox Virus A29L (MPXV/USA/2022/MA001) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Monkeypox Virus A29L is a gene that is found in the genome of the Monkeypox virus (MPXV). MPXV is a member of the Orthopoxvirus genus, which also includes the variola virus (which causes smallpox) and the vaccinia virus (used in smallpox vaccination). The A29L gene encodes a protein that is involved in various aspects of the viral life cycle, including viral replication, assembly, and immune evasion.
Spezifikation
| Monkeypox Virus A29L (MPXV/USA/2022/MA001) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2527 | |
| Unconjugated | |
| A29L | |
| Full length of Monkeypox virus A29L recombinant protein (MPXV_USA_2022_MA001). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur