Learn More
Monomethylsuberat, TRC
Specifications
Specifications
| Chemischer Name oder Material | Monomethyl Suberate |
| CAS | 3946-32-5 |
| Summenformel | C9H16O4 |
| Menge | 1 g |
| Synonym | Octanedioic Acid, Monomethyl Ester, Suberic Acid Methyl Ester, Suberic Acid, Monomethyl Ester, 8-Methoxy-8-oxooctanoic Acid, Methyl 1,8-Octanedionate, Methyl 7-Carboxyheptanoate |
| InChI-Formel | InChI=1S/C9H16O4/c1-13-9(12)7-5-3-2-4-6-8(10)11/h2-7H2,1H3,(H,10,11) |
| SMILES | COC(=O)CCCCCCC(=O)O |
| IUPAC-Name | 8-Methoxy-8-Oxooctansäure |
| Molekulargewicht (g/mol) | 188.22 |
| Formelmasse | 188.1049 |