Learn More
Invitrogen™ Motavizumab Humanized Recombinant Human Monoclonal Antibody
Beschreibung
Motavizumab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Motavizumab is a second generation, ultra-potent, affinity-matured, humanized mAb derived from palivizumab that reduces replication of Respiratory Syncytial Virus (RSV), a pathogen which causes lower respiratory tract disease.
Spezifikation
Spezifikation
| Antigen | Motavizumab Humanized |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Gen-Alias | MEDI-524 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Mehr anzeigen |