Learn More
CSF1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CSF1.
Marke: Abnova H00001435-A01.50uL
Beschreibung
The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of macrophages. The active form of the protein is found extracellularly as a disulfide-linked homodimer, and is thought to be produced by proteolytic cleavage of membrane-bound precursors. The encoded protein may be involved in development of the placenta. Four transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Sequence: EEVSEYCSHMIGSGHLQSLQRLIDSQMETSCQITFEFVDQEQLKDPVCYLKKAFLLVQDIMEDTMRFRDNTPNAIAIVQLQELSLRLKSCFTKDYEEHDKSpezifikation
| CSF1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CSF1. | |
| CSF1 | |
| MCSF/MGC31930 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC021117 | |
| CSF1 | |
| CSF1 (AAH21117, 33 a.a. to 132 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 1435 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only