Learn More
Mouse anti-Rat IgA Heavy Chain Secondary Antibody (MARA-1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-68392IR
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Spezifikation
| IgA Heavy Chain | |
| Monoclonal | |
| DyLight 755 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| MARA-1 | |
| 50mM Sodium Borate | |
| Purified IR1060 IgA rat myeloma protein | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |