missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marke: R&D Systems AF6325-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BMP-6 Polyclonal specifically detects BMP-6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Spezifikation
| BMP-6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, Vgr-1, VGR1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 654 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P20722 | |
| BMP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse BMP-6 Ser372-His510 Accession # P20722 | |
| 25 μg | |
| Primary | |
| Detects recombinant human and mouse BMP-6 in direct ELISAs and detects mouse BMP-6 in Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) BMP-5 and rmBMP-7 is observed. | |
| Mouse | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur