Learn More
Invitrogen™ Mouse Cardiac Troponin I (TNNI3) ELISA Kit
Click to view available options
Menge:
96 Tests
Packungsgröße:
96 Stück
Beschreibung
Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).
Spezifikation
Spezifikation
| Zugriffsnummer | P48787 |
| Assay-Bereich | 7,81 bis 500 pg/ml |
| Assay-Empfindlichkeit: | 4.69 pg/mL |
| Konjugat | HRP |
| Produkttyp | ELISA |
| Probentyp | Serum |
| Zur Verwendung mit (Geräte) | Kolorimetrisches Mikrotiterplatten-Lesegerät |
| Gen-ID (Entrez) | 21954 |
| Gensymbol | TNNI3 |
| Interassay-VK | <10% |
| Mehr anzeigen |