missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CLEC3B/Tetranectin (NP_035736) VersaClone cDNA
Beskrivelse
- The Mouse CLEC3B/Tetranectin (NP_035736) VersaClone cDNA clone contains the complete ORF for CLEC3B/Tetranectin, along with a Kozak consensus sequence for optimal translation initiation
- The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
- The CLEC3B/Tetranectin insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Tekniske data
Tekniske data
| Gen | mCLEC3B/Tetranectin |
| Zugriffsnummer | NP_035736 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Gen-ID (Entrez) | 7123 |
| Inhalt und Lagerung | Store the unopened product at -20 to -80°C |
| Spezies | Mouse |
| Menge | 10 μg |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?