Learn More
R&D Systems™ Mouse Coagulation Factor II (NP_034298.1) VersaClone cDNA
Artikelnummer. 18789996
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18789996 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18789996 Lieferant R&D Systems™ Lieferanten-Nr. RDC3237
  • ONLINE EXCLUSIVE PRICE
    761.00 €
    719.25 € / 10 Mikrogramm
    Sparen 41.75 €
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 20-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Spezifikation
Gen Coagulation Factor II/Thrombin
Zugriffsnummer NP_034298.1
Gen-ID (Entrez) 2147
Gen-Alias Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), F2, PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin, Thrombin receptor-like 2
Inhalt und Lagerung Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Spezies Mouse
Menge 10 μg