missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EDAR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
211.00 € - 523.00 €
Spezifikation
| Antigen | EDAR |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
| Anwendungen | Western Blot, Blockade of Receptor-ligand Interaction |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18741158
![]() |
R&D Systems
AF745-SP |
25 μg |
211.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18022592
![]() |
R&D Systems
AF745 |
100 μg |
523.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
EDAR Polyclonal specifically detects EDAR in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Spezifikation
| EDAR | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Mouse | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| EDAR | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Q9R187 | |
| 10913 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ala187 Accession # Q9R187 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse EDAR in direct ELISAs and Western blots. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts