Learn More
Mouse Erythropoietin/EPO Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18761797
Change view
Click to view available options
Menge:
100 μg
25 μg
500 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
18761797 500 μg 500 Mikrogramm
18716729 25 μg 25 Mikrogramm
18740928 100 μg 100 Mikrogramm
3 options
Artikelnummer. 18761797
Marke: R&D Systems MAB9591500
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rat Monoclonal Antibody
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Erythropoietin/EPO
Anwendungen Neutralization, ELISA
Klassifikation Monoclonal
Klon 148436
Konjugat Unconjugated
Verdünnung Neutralization 0.15-0.75 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer P07321
Gen-Alias ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Gensymbole EPO
Wirtsspezies Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Erythropoietin/EPO, Ala27-Arg192, Accession # P07321
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 500 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 2056
Testspezifität Detects mouse Erythropoietin/EPO in direct ELISAs. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant rat Erythropoietin/EPO, 5-15% cross-reactivity with recombinant human Erythropoietin/EPO, and no cross-reactivity with recombinant mouse Thrombopoietin is observed.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Mouse
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a