missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GITR Ligand/TNFSF18 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Marke: R&D Systems FAB21773R-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry
Spezifikation
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 994529 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7TS55 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8995 | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur