Learn More
Invitrogen™ Mouse IgG kappa Humanized Recombinant Human Monoclonal Antibody (SAA0490)
Artikelnummer. 30131002
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
30131002 100 μg 100 Mikrogramm
1 options
Artikelnummer. 30131002
Marke: Invitrogen™ MA554746
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Human Recombinant Monoclonal Antibody
Compatible with Direct ELISA.
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Mouse IgG kappa Humanized
Anwendungen ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon SAA0490
Konzentration 2.63 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen-Alias IgG1; Igh-4; Ighg1; Immunoglobulin G; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; VH7183
Wirtsspezies Human
Immunogen Recombinant Mouse Kappa chain.
Reinigungsverfahren Protein A/G
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype VHH-hFc