missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-15 Recombinant Protein, PeproTech®

Artikelnummer. 17870213
Click to view available options
Menge:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
2 Stück
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17870213

Marke: Gibco™ 21015100UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

210-15-1MG will be provided as 2 x 500 μg (210-15-500UG). Recombinant Murine IL-15 is a 13.3 kDa protein consisting of 115 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P48346
Zur Verwendung mit (Anwendung) Control, Western Blot Control, ELISA Standard, Bioactivity
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 16168
Molekulargewicht 13.3 kDa
Name Mouse IL-15
Reinigungsverfahren Purified
Menge 100 μg
Quelle E. coli
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/μg
Gen-Alias AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Gebräuchliche Bezeichnung IL-15
Gensymbol IL15
Biologische Aktivität The ED50 was determined by the stimulation of the proliferation of murine CTLL-2 cells is ≤ 5.0 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS
Inhalt und Lagerung -20°C
Aktivität The ED50 was determined by the stimulation of the proliferation of murine CTLL-2 cells is ≤ 5.0 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg.
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt