Learn More
Mouse IL-15R alpha Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Marke:  R&D Systems AF551
Artikelnummer. 18070102
  • ONLINE EXCLUSIVE PRICE
    497.00 €
    469.35 € / 100 Mikrogramm
    Sparen 27.65 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
IL-15R alpha Polyclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IL-15R alpha
Polyclonal
Unconjugated
Q60819
IL15RA
Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha Gly33-Lys205 Accession # Q60819
100 μg
Primary
Detects mouse IL-15 R alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IL-15 R alpha is observed and less than 1% cross-reactivity with rhIL-2 R alpha , recombinant mouse (rm) IL-2 R beta , and rmIL-2 R gamma is observed.
Mouse
IgG
Western Blot, Flow Cytometry, Neutralization, CyTOF
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.3-1.5 ug/mL, CyTOF-ready
CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Goat
Affinity Purified
RUO
3601
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen