missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-21, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
Beschreibung
Recombinant Murine IL-21 is a 15.0 kDa protein consisting of 130 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
Spezifikation
Spezifikation
| Zugriffsnummer | Q9ES17 |
| Zur Verwendung mit (Anwendung) | Functional Assay |
| Zusammensetzung | protein with no preservative |
| Gen-ID (Entrez) | 60505 |
| Molekulargewicht | 15 kDa |
| Name | Mouse IL-21, Animal-Free |
| Menge | 100 μg |
| Quelle | E. coli |
| Kennzeichnung | RUO |
| Endotoxin-Konzentration | <0.1 EU/ μg |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur