missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-9 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems AB-409-NA
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-9 Polyclonal specifically detects IL-9 in Mouse samples. It is validated for Western Blot, Neutralization.
Spezifikation
| IL-9 | |
| Polyclonal | |
| Unconjugated | |
| P15247 | |
| IL9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-9 Gln19-Pro144 Accession # P15247 | |
| 1 mg | |
| Primary | |
| Detects mouse IL-9 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human IL-9 is observed. Neutralizes the biological activity of recombinant mouse IL-9. It will not neutralize the biological activity of recombinant human IL-9. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 3-18 ug/mL | |
| Cytokine P40, HP40, IL9, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3578 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur