missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-10 Recombinant Protein

Artikelnummer. 11868791
Click to view available options
Menge:
5 μg
Packungsgröße:
5 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 11868791

Marke: Invitrogen™ RMIL105

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Recombinant Protein

Use: Mouse IL-10 exerts activity in the range of 0.2 to 20 ng/mL for most in vitro applications. Reconstitution and Dilution: Reconstitute in distilled water to a concentration of 100 μg/mL. After reconstitution, dilute cytokine in a neutral buffer containing carrier protein such as 5-10% FBS or 1-4% high purity BSA (fatty acid free fraction V). Lyophilized product is stable until expiration if stored desiccated below 0°C. Cytokines diluted as indicated above should be divided into aliquots and stored at -70°C. Recombinant proteins should be stored at concentrations >500 ng/mL to reduce loss of activity.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P18893
Zur Verwendung mit (Anwendung) Control, ELISA, Immunoprecipitation, Western Blot
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 16153
Molekulargewicht 18.7 kDa
Name Mouse IL-10
Menge 5 μg
Quelle E. Coli
Kennzeichnung RUO
Gen-Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Gebräuchliche Bezeichnung IL-10
Gensymbol Il10
Biologische Aktivität ED50 ≤ 2 ng/mL; determined by the dose-dependent proliferation of human IL-4 co-stimulated MC/9 cells.
Konjugat Unconjugated
Spezies E. coli
Rekombinant Recombinant
Sequenz Mouse IL-10 recombinant protein contains 161 amino acids
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Lyophilized
Protein-Subtyp Recombinant
Reinheits- oder Qualitätsgrad >98% by SDS-PAGE and HPLC
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt