missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Klotho beta Biotinylated Antibody, R&D Systems™
Beschreibung
Klotho beta Polyclonal specifically detects Klotho beta in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Klotho beta |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | Biotin |
| Verdünnung | Western Blot 0.1 ug/mL |
| Gen-Zugriffsnummer | NP_112457 |
| Gen-Alias | beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 |
| Gensymbole | KLB |
| Wirtsspezies | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?