missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Laminin-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB2549-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Laminin-1 Monoclonal specifically detects Laminin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Adhesion Blockade.
Spezifikation
| Laminin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284217 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Adhesion Blockade | |
| AL-2 | |
| Western Blot, Immunohistochemistry 8-25 ug/mL, Adhesion Blockade | |
| LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha | |
| Purified fragment of chymotrypsin-digested mouse EHS tumor-derived Laminin-1 | |
| 25 μg | |
| Primary | |
| Reacts with mouse Laminin-1 A and B chains on the long arm below the cross region. | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur