missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lipocalin-2/NGAL Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Marke: R&D Systems MAB1857-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for Western Blot.
Spezifikation
| Lipocalin-2/NGAL | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3934 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 228418 | |
| Western Blot 0.5 ug/mL | |
| P11672 | |
| LCN2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Lipocalin-2 aa 21-200 Accession # P11672 | |
| 25 μg | |
| Primary | |
| Detects recombinant mouse (rm)Lipocalin-2 in direct ELISAs and Western blots. In these formats, does not cross-react with rhLipocalin-1 or rhLipocalin-2. | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur