missing translation for 'onlineSavingsMsg'
Learn More

Mouse Mer PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18727688
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
18727688 100 μg 100 Mikrogramm
1 options
This item is not returnable. View return policy

Product Code. 18727688

Brand: R&D Systems FAB591P100UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Mer Polyclonal specifically detects Mer in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mer
Anwendungen Flow Cytometry
Klassifikation Polyclonal
Konjugat R-PE
Verdünnung Flow Cytometry 0.1 ug/10^6 cells
Zusammensetzung Supplied in a saline solution containing BSA and Sodium Azide.
Gen-Zugriffsnummer Q60805
Gen-Alias c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Wirtsspezies Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer Glu23-Phe498 Accession # Q60805
Reinigungsverfahren Affinity Purified
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10461
Testspezifität Detects mouse Mer in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human Mer is observed and less than 1% cross-reactivity with recombinant mouse (rm) Axl and rmDtk is observed.
Zielspezies Mouse
Inhalt und Lagerung Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.