missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MGMT Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB3299-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MGMT Monoclonal specifically detects MGMT in Mouse samples. It is validated for Western Blot.
Spezifikation
| MGMT | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P26187 | |
| MGMT | |
| E. coli-derived recombinant mouse MGMT Met1-Asn211 Accession # P26187 | |
| 25 μg | |
| Primary | |
| Detects mouse MGMT in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 300008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4255 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur