missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse NgR2/NgRH1 (NP_954693) VersaClone cDNA

Product Code. 18790798
Change view
Click to view available options
Menge:
10 μg
Unit Size:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
18790798 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18790798 Supplier R&D Systems™ Supplier No. RDC2508
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10 Mikrogramm
    Sparen 18.25 €
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Mouse NgR2/NgRH1 (NP_954693) VersaClone cDNA clone contains the complete ORF for NgR2/NgRH1/RTN4RL2, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The NgR2/NgRH1/RTN4RL2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gen mNgR2/NgRH1
Zugriffsnummer NP_954693
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-ID (Entrez) 349667
Inhalt und Lagerung Store the unopened product at -20 to -80°C
Spezies Mouse
Menge 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.