Learn More
Mouse SR-AI/MSR Antibody, R&D Systems™
GREENER_CHOICE
Artikelnummer. 18189520
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18189520 100 μg 100 Mikrogramm
18783947 25 μg 25 Mikrogramm
2 options
Artikelnummer. 18189520
Marke: R&D Systems MAB1797
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rat Monoclonal Antibody has been used in 2 publications
SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SR-AI/MSR
Anwendungen Western Blot, Flow Cytometry, CyTOF
Klassifikation Monoclonal
Klon 268318
Konzentration LYOPH
Konjugat Unconjugated
Verdünnung Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gen-Zugriffsnummer AAA39747
Gen-Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI
Gensymbole MSR1
Wirtsspezies Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SR-AI/MSR Trp79-Ser454 Accession # AAA39747
Reinigungsverfahren Protein A or G purified from hybridoma culture supernatant
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 4481
Testspezifität Detects mouse SR-AI/MSR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SR-AI is observed.
Rekonstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Zielspezies Mouse
Inhalt und Lagerung Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b