Learn More
Mouse TGF-beta RI/ALK-5 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marke:  R&D Systems MAB587
Artikelnummer. 18103261
  • ONLINE EXCLUSIVE PRICE
    537.00 €
    507.15 € / 500 Mikrogramm
    Sparen 29.85 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TGF-beta RI/ALK-5
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
7046
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Western Blot
141229
Western Blot 1 ug/mL
BAA05023
TGFBR1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023
500 μg
Primary
Detects mouse TGF-beta RI/ALK-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 20-25% cross-reactivity with recombinant human (rh) TGF-beta RI and no cross-reactivity with recombinant rat MIS RII, rhTGF-beta RII, rhTGF-beta RIIb, or rhTGF-beta RIII.
Mouse
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen