missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TIM-5 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems BAF3374
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TIM-5 Polyclonal specifically detects TIM-5 in Mouse samples. It is validated for Western Blot.
Spezifikation
| TIM-5 | |
| Polyclonal | |
| Biotin | |
| NP_001156829 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 210535 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Gm12169 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 Accession # NP_001156829 | |
| 50 μg | |
| Primary | |
| Detects mouse TIM-5 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-6 is observed and less than 2% cross-reactivity with rmTIM-2, rmTIM-3, and rmTIM-4 is observed. | |
| Mouse | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur