missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse VEGF-165, AOF Recombinant Protein

Artikelnummer. 17676565
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17676565

Marke: Invitrogen™ RP87758

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

E. coli Recombinant Protein

The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 165, 230 aa.
  • Biological Activity: <10 ng/mL
  • Molecular Weight: 19.4, 38.8 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q00731
Zur Verwendung mit (Anwendung) Control, ELISA, Functional Assay, Gel Shift, Western Blot
Zusammensetzung 0.1% TFA with no preservative
Gen-ID (Entrez) 22339
Molekulargewicht 19.4, 38.8 kDa
Name Mouse VEGF-165, AOF
Menge 100 μg
Quelle E. coli
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Gen-Alias H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165 b; Vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; Vascular permeability factor; VEGF; VEGF165; Vegfa; VEGF-A; VPF
Gensymbol Vegfa
Spezifische Reaktivität Mouse
Konjugat Unconjugated
Rekombinant Recombinant
Form Lyophilized
Reinheits- oder Qualitätsgrad >95%
Mehr anzeigen Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.