missing translation for 'onlineSavingsMsg'
Learn More

MPZL1 Antibody, Novus Biologicals™

Artikelnummer. 18602507 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
25 μL
Packungsgröße:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18602507 25 μL 25 Mikroliter
18244373 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18602507 Lieferant Novus Biologicals Lieferanten-Nr. NBP25748725ul

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

MPZL1 Polyclonal specifically detects MPZL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen MPZL1
Anwendungen Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Zusammensetzung PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gen-Alias FLJ21047, immunoglobulin family transmembrane protein, MPZL1b, myelin protein zero-like 1, myelin protein zero-like protein 1, protein zero related, Protein zero-related, PZR1b, PZRb, PZRPZRa
Gensymbole MPZL1
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NSKRDYTGCSTSESLSPVKQAPRKSPSDTEGLVKSLPSGSHQGPVIYAQLDHSGGHHSDKINKSESVVYADIRKN
Reinigungsverfahren Affinity Purified
Menge 25 μL
Regulatorischer Status RUO
Forschungsgebiet Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 9019
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.